Learn More
Thermo Scientific™ Orion™ Ionplus™ Sure-Flow™ Lead Combination Ion Selective Electrode
Click to view available options
Connector Types:
BNC
Description
Measure lead ions in aqueous solutions quickly, simply, accurately and economically with Thermo Scientific™ Orion™ Lead Electrodes.
- Free-flowing Themo Scientific™ Sure-Flow™ junction assures stable, drift-free potentials
- When conventional electrode junctions clog, the Sure-Flow junction can be opened and flushed clean simply by pressing the cap
- Lead Half-Cell Electrode used in conjunction with the Orion 900200 Double Junction Reference electrode offers the benefits of the Sure-Flow™ junction design
- Calibration standards catalog number: 948206 - 0.1 M Pb(CIO4)2; 948207 - 0.1 M Na2SO4
- Required ISA Catalog numbers: Consult instruction manual
Specifications
Specifications
Connector Types | BNC |
Description | With BNC connector |
Drift | drift-free potentials |
Junction | Sure-Flow Junction |
For Use With (Equipment) | ISE Meter |
Type | Lead Ion Selective Electrode, BNC connector |