Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Thermo Scientific™ Orion™ Single Junction Spear Tip Glass Bodied Combination pH Electrode
Click to view available options
Connector Types:
BNC
Description
Thermo Scientific™ Orion™ Spear Tip pH Electrodes provide reliable and accurate measurements of routine samples. Best suited for piercing solid/semi-solid samples and measuring small volume samples.
Orion™ Single Junction Electrodes provide a reliable option for routine pH measurement. These silver chloride electrodes are available in a variety of styles and provide accurate measurements of typical samples.
Solutions required
Electrode fill solution
900011 4M KCl with Ag/AgCl filling solution, 5 x 60 mL bottles
Electrode storage solution
910001 pH electrode storage solution, 475 mL bottle
pH buffers
910168 pH 1.68 buffer, NIST traceable, 475 mL
910104 pH 4.01 buffer, color coded red, NIST traceable, 475 mL
910105 pH 5.00 buffer, color coded orange, NIST traceable, 475 mL
910686 pH 6.86 buffer, DIN standard, NIST traceable, 475 mL
910107 pH 7.00 buffer, color coded yellow, NIST traceable, 475 mL
910918 pH 9.18 buffer, DIN standard, NIST traceable, 475 mL
910110 pH 10.01 buffer, color coded blue, NIST traceable, 475 mL
910112 pH 12.46 buffer, NIST traceable, 475 mL
910199 Standard All-in-One pH buffer kit, includes pH 4.01, 7.00 and 10.01 buffers, 475ml each, pH
electrode storage solution, 475ml, and 12 mm electrode storage bottle
electrode storage solution, 475ml, and 12 mm electrode storage bottle
pH cleaning solution kit
900020 Cleaning solution A and C, 30 ml each. B and D, 60 ml each
Items included with
900011 4M KCl with Ag/AgCl filling solution 60mL 1 bottle
Warranty period
12 months from date of purchase
Specifications
Specifications
| Description | With BNC waterproof connector |
| Connector Types | BNC |
| Material | Glass Body |
| pH Range | 0 to 14 |
| Temperature (Metric) Operating | 0°C to 90°C |
| Type | Half-Cell Electrode, low resistance, BNC connector |
| For Use With (Equipment) | Orion pH Meter |
| Internal Reference | Ag/AgCl |
| Precision | 0.02 |
| Length (Metric) | 110 mm |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction