Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Thermo Scientific™ Orion™ ISE Surfactant Analysis Solutions
Click to view available options
Quantity:
60 mL
475 mL
Chemical Name or Material:
Hyamine 1622 Titrant
Sodium Lauryl Sulfate Standard
Surfactant Reagent, Nonionic
Description
Use Thermo Scientific™ Orion™ ISE Surfactant Analysis Solutions for titrations with surfactant ion selective electrodes (ISE).
All surfactant analysis solutions are packaged as one 1 pint (475mL) bottle, except Cat. No. 654202 packaged as one 2 oz. (60mL) bottle.
Specifications
Specifications
| Chemical Name or Material | Surfactant Reagent, Nonionic |
| Color | Blue |
| Description | 654205 |
| For Use With (Equipment) | Surfactant Ion Selective Electrode |
| Physical Form | Liquid |
| Quantity | 475 mL |
| Solution Type | Surfactant Analysis |
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction