Learn More
Thermo Scientific™ Orion™ Powder Chemistry Packs
Catalog No. 13642221
Click to view available options
Type:
Chlorine (free)
Chlorine (total)
Copper (free)
Iron (ferro)
Iron (total)
Manganese
Molybdate/Molybdenum
Nitrite
Phosphate (ortho)
Silica
Sulfate
Range:
0.01 to 0.3 mg/L N
0.02 to 1.8 mg/L Fe
0.02 to 2 mg/L Cl2
0.02 to 3 mg/L Fe
0.05 to 5 mg/L Cu
0.06 to 2.5 mg/L PO4
0.1 to 18 mg/L Mn
0.5 to 66 mg/L MoO4
1 to 90 mg/L SiO2
5 to 100 mg/L SO4
Achieve easier and cleaner measurements with pre-measured and ready-to-use powder chemistry packs.
Use Thermo Scientific™ Orion™ AQUAfast Powder Pack Reagents with Orion AQUAfast colorimeters and Thermo Scientific Orion AquaMate Vis and UV-Vis spectrophotometers. Powder pack chemistries dissolve quickly in solution, are easy to mix and do not require a stir rod or crushing. Each powder pack is notched for simplified pouch opening in the lab or field. Powder pack reagents measure aluminum, ammonia, chlorine, copper, cyanide, DEHA, iron, manganese, molybdate, nitrite, phosphate, silica and sulfate.
  • Equivalent to other widely used and recognized chemistries
  • Compatible with Orion™ AQUAfast™ Colorimeters
TRUSTED_SUSTAINABILITY
Specifications
Type Copper (free)
EPA Approved Methods Wastewater: Method 8506
For Use With (Equipment) Orion AquaMate Vis and UV-Vis Spectrophotometers, Orion AQUAfast AQ4000 Colorimeter, Orion AQUAfast AQ3700 Colorimeter
Includes 100 Cu 1 Powder Packs
Packaging Powder Pack
Quantity 100 tests
Range 0.05 to 5 mg/L Cu
Recommended Accessories AQUAfast 24 mm round vials (Cat. No. AC2V24)
Test Method Bicinchoninate
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.