Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Orion™ ROSS™ Combination Flat Surface pH Electrodes PROMO

Catalog No. p-7230521
Click to view available options
Connector Types:
BNC

When you purchase a Thermo Scientific™ ROSS™ pH Electrode, you’ll receive a ROSS Buffer Kit at no additional cost. Order using the catalog numbers below and your products will ship together.

The 8135BN ROSS Combination Flat Surface pH Electrode measures pH of soft, moist surfaces — both solid and semi-solid —such as agar gel plates, meats, bread dough and similar samples. With drift less than 0.002 pH per day and minimized restandardization, this electrode works in extreme cases of samples varying from one another by 50°C or more.

  • Material: Epoxy body
  • pH range: 0 to 14
  • Temperature range: 0 to 100°C
  • Internal reference: ROSS
  • Junction: Glass fiber
  • Dimensions: 120 × 12 mm
  • pH precision: 0.01
  • Fill solution: Orion 810007
  • Cap diameter: 16 mm
  • Small samples are available in micro sample dish
  • Flat surface tip

Includes: One 60 mL bottle of ROSS fill Solution, 810007 and spout cap

TRUSTED_SUSTAINABILITY

Specifications

Description With BNC connector
Connector Types BNC
Junction Glass Fiber
Material Epoxy Body
pH Range 0 to 14
Temperature (Metric) Operating 0°C to 100°C
Type Combination Flat Surface pH Electrode, BNC Connector
Drift <0.002 pH/day
Filling Solution Type 3M KCl
For Use With (Equipment) Standard pH Meter
Internal Reference ROSS
Precision 0.01
Storage Requirements Short-term Storage (up to one week): Soak the electrode in ROSS' pH electrode storage solution, Cat. No. 810001, or standard pH electrode storage solution, Cat. No. 910001. For enhanced electrode performance, use ROSS' pH electrode storage solution. Long-term Storage (more than one week): Fill the reference chamber and securely cover the fill hole. Cover the sensing bulb and reference junction with the protective cap, sleeve or storage bottle containing a few drops of storage solution. Before returning the electrode to use, prepare it as a new electrode.
Temperature (English) Operating 32 to 212°F
Length (English) Cable 39 in.
Length (Metric) Cable 1 m
Diameter (English) Body 0.47 in.
Diameter (Metric) Body 12 mm
Diameter (Metric) Cap 16 mm
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.