Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Thermo Scientific™ Orion™ ROSS™ Combination pH Electrodes PROMO
Click to view available options
Connector Types:
BNC
Description
Features:
- Works in extreme case of samples varying from one another by 50°C or more
- Drift is less than 0.002 pH per day, restandardization is minimized
- General purpose, top performance combination electrode
- Double junction
- TRIS compatible
- Waterproof BNC connector
Includes:
- One bottle 810007 fill solution
Requires:
- pH buffers for calibration
- Beakers, plastic or glass
- Magnetic stirrer or Orion stirrer probe
- Distilled or deionized water
Recommended for:
- Quality Control (QC) and research applications

Specifications
Specifications
Description | BNC connector |
Connector Types | BNC |
Junction | Ceramic |
Material | Glass Body |
pH Range | 0 to 14 |
Temperature (Metric) Operating | 0°C to 100°C |
Type | Combination pH Electrode, BNC Connector |
Drift | <0.002 pH/day |
Filling Solution Type | 3M KCl |
For Use With (Equipment) | Standard pH Meter |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction