Learn More
Thermo Scientific™ Orion™ ROSS Ultra™ Low Maintenance pH/ATC Triode™ Combination Electrodes PROMO
Description
Measure pH and Temperature with One Low-Maintenance Electrode
Low ionic strength samples, such as clean waters, present a challenge to pH electrode performance. Sluggish response, poor accuracy and drift are common. The Thermo Scientific Orion ROSS Ultra 8107 Triode pH electrode responds within one minute, proving a stable, accurate reading.
Complete your pH measurements faster. Get long, maintenance-free electrode lifetime with special long-life gel and innovative capillary glass junction design. Maintain sample integrity, even in TRIS buffers; ROSS double junction design electrodes contain no silver, which can bind with biological samples. With built-in temperature sensors, these electrodes offer a convenient solution for your routine sample measurements.
Compatible with Thermo Scientific™ Orion™ Versa Star™, Thermo Scientific™ Orion Star™ A, and Thermo Scientific™ Orion™ Dual Star™ meters, as well as Mettler Toledo™ pH meters.
Target Markets:
- Pharmaceutical/Biopharmaceutical
- Food Manufacturing
- Academic/Government
- Environmental
- Quality Control
- Research
- General Purpose
Specifications
Specifications
| Description | Orion ROSS Ultra Low Maintenance pH/ATC Triode Combination Electrodes with a BNC and RCA Connector and 1 meter cable |
| Connector Types | BNC, RCA |
| Junction | Glass Capillary |
| Material | Epoxy Body |
| pH Range | 0 to 14 |
| Temperature (Metric) Operating | 0°C to 80°C |
| Type | pH/ATC Combination Electrode |
| Filling Solution Type | None required |
| For Use With (Equipment) | Mettler Toledo 5/7 Series pH Meters |
| Internal Reference | ROSS |
| Show More |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.