Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Orion™ ROSS Ultra™ Epoxy Bodied pH/ATC Triode™

Catalog No. p-4529932
Click to view available options
Connector Types:
BNC (Waterproof), 8-pin Mini-DIN
BNC (Waterproof), RCA
Length (Metric) Cable:
1 m
3 m

Catalog No. 13-641-981

Supplier: Thermo Scientific™ 8107BNUMD

Only null left
Add to Cart
Add to Cart

Accurately measure pH and temperature together in just one electrode, with a built-in temperature sensor.

Thermo Scientific™ Orion™ ROSS Ultra Triode™ 3-in-1 pH/Automatic Temperature Compensation Electrodes combine the precision and speed of response of the ROSS Ultra line with a built-in ATC. These products offer a low-maintenance, gel-filled, epoxy body and are best used where minimal maintenance is required. Non-refillable Orion ROSS Ultra Triodes come with an 18 month warranty.
 

Measure pH and Temperature with One Low-Maintenance Electrode

Low ionic strength samples, such as clean waters, present a challenge to pH electrode performance. Sluggish response, poor accuracy and drift are common. The Thermo Scientific Orion ROSS Ultra 8107 Triode pH electrode responds within one minute, proving a stable, accurate reading.

Complete your pH measurements faster. Get long, maintenance-free electrode lifetime with special long-life gel and innovative capillary glass junction design. Maintain sample integrity, even in TRIS buffers; ROSS double junction design electrodes contain no silver, which can bind with biological samples. With built-in temperature sensors, these electrodes offer a convenient solution for your routine sample measurements.

Compatible with Thermo Scientific™ Orion™ Versa Star™, Thermo Scientific™ Orion Star™ A, and Thermo Scientific™ Orion™ Dual Star™ meters, as well as Mettler Toledo™ pH meters.

Target Markets:

  • Pharmaceutical/Biopharmaceutical
  • Food Manufacturing
  • Academic/Government
  • Environmental
Applications:
  • Quality Control
  • Research
  • General Purpose
TRUSTED_SUSTAINABILITY

Specifications

Description Orion ROSS Ultra Low Maintenance pH/ATC Triode Combination Electrodes with a BNC and 8-pin MiniDIN Connector and 1 meter cable
Connector Types BNC (Waterproof), 8-pin Mini-DIN
Junction Glass Capillary
Material Rugged Epoxy Body, Filled with Low Maintenance Gel
pH Range 0 to 14
Temperature (Metric) Operating 0°C to 80°C
Type pH/ATC Combination Electrode
Filling Solution Type None required
For Use With (Equipment) Orion Versa Star, Star A and Dual Star Meters
Internal Reference ROSS
Isopotential Point 7
pH Relative Accuracy 0.03 pH for buffers using automatic temperature compensation
Precision 0.01
Sample Compatibility TRIS, sulfides, or proteins
Warranty 18 Months
Length (Metric) 120 mm
Length (Metric) Cable 1 m
Diameter (Metric) 12 mm
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.