Learn More
Thermo Scientific™ Orion Star™ A216 pH/Dissolved Oxygen Benchtop Multiparameter Meter
Reliably measure pH, dissolved oxygen and temperature with the Thermo Scientific™ Orion™ Star A216 pH / DO / RDO Benchtop Meter for advanced lab analysis and interfacing
Manufacturer:  Thermo Scientific™ STARA2160
 View more versions of this product
Catalog No. 13-645-548
  Check Availability
Add to cart
Specifications
Description
Record accurate and reliable pH, mV, ORP, dissolved oxygen and temperature measurements with the Thermo Scientific™ Orion™ Star A216 pH, DO and RDO Benchtop Meter, ideal for a wide range of applications and advanced pH and dissolved oxygen analysis in the lab. Use the two channels to measure pH and dissolved oxygen simultaneously or view each channel separately. Use an RDO optical or polarographic DO probe for ultimate flexibility in sensors. Log up to 2000 data point sets with time / date stamp and easily transfer calibration and data logs via USB or RS232 to a printer or computer.
The Thermo Scientific Orion Star A216 pH/ RDO/Dissolved Oxygen Benchtop Meter is the ideal choice for any lab. Get the information you need quickly and easily from the large, backlit graphic LCD display. Informative display shows individual pH/mV/RmV result, dissolved oxygen in percent saturation or concentration results with temperature, or all results at once. Additional helpful information includes electrode status, time, date, sample ID, user ID and calibration. For advanced users, features such as stability and averaging choices provide additional options.
  • Anyone can use this meter with its plain language screen prompts, soft keys that update for easy selection and a multilanguage interface that supports English, Spanish, French, Italian, German and Chinese
  • Don't miss a reading — AUTO-READ™ locks in the stable reading on your screen, ready indicator alerts when readings are stable and timed reading gathers data in specific time intervals
  • Up to five point pH calibration with automatic recognition for USA/NIST™ and DIN buffers and calibration editing to fix errors without a complete recalibration
  • No need to worry about DO probe compatibility — the meter accepts and automatically recognizes Orion polarographic and RDO optical/luminescence-based probes
  • Accurate DO results are easier with automatic temperature, manual salinity and automatic/ manual barometric pressure compensation
  • Calibrate DO by using water-saturated air, air-saturated water, a Winkler titration or zero point calibration
  • Nonvolatile memory holds up to 2000 data points with time and date stamp
  • Easily transfer data and keep meter software up to date with the USB and RS232 ports and complimentary data analysis software
  • To mix samples without a stir plate or for faster BOD readings, meter can directly control the 096019 Orion Star stirrer or 086030MD Orion AUTO-STIR™ DO probes (each sold separately)
  • Included electrode arm and newly designed probe holder make it easier to maintain and place probes into samples
  • Works with almost every AC power source with the included universal power adapter or with four AA batteries (sold separately) to run the meter on DC power
  • IP54-rated housing handles splashes and is wall-mountable if table space is a problem
  • Three year meter replacement warranty
Description & Specifications
Specifications
Orion Star A216 pH/RDO/DO benchtop meter with stand
Orion Star A216 pH/RDO/DO benchtop meter with stand
-2.000 to 20.000pH
±0.002pH
±2000.0mV
0.1mV
7.000pH
0.01, 0.1mg/L
0.00 to 50.00mg/L
±0.1mg/L up to 8mg/L, ±0.2mg/L from 8 to 20mg/L, ±10% of reading up to 50mg/L
0.1, 1 % saturation
0.0 to 500.0 % saturation
±2 % saturation ? 200 % saturation, ±10 % saturation > 200 % saturation
400 to 850mmHg
±6mmHg
pH with calibration editing option, relative mV (RmV), ORP, dissolved oxygen and temperature
1 point relative mV (RmV) mode, 1 point ORP mode
-5.0° to 105.0° C, 22.0° to 221.0° F
±0.1°C
1 point temperature offset calibration
Graphic LCD with backlight
Comprehensive with menu-specific function keys and dual purpose scroll/shortcut keys
Selectable fast, medium, slow or Smart Stability setting; Selectable off or Automatic Smart averaging setting
2000 with date and time stamp
Non-volatile memory preserves data log, calibration log and meter settings
10 per channel with password protection
BNC (pH or ORP electrode), pin-tip (reference electrode), 8 pin MiniDIN (ATC temperature probe), 9 pin MiniDIN (RDO optical or polarographic DO sensor with built-in temperature), stirrer input (stirrer probe)
Universal AC adapter (included) or 4 AA batteries (sold separately)
Four AA (sold separately)
IP-54
41° to 113°F (ambient)
5 to 85 %, non-condensing
0.9kg
Orion Star A216 pH/mV/RmV/ORP/RDO Optical/Dissolved Oxygen/Temperature Benchtop Meter
3 year meter warranty
Star A216 pH/RDO/DO meter, electrode stand, universal power adapter, literature CD, printed quick start guide, computer interface cable and meter test certificate
0.1, 0.01, 0.001pH
±2000.0mV
±2000.0mV
±0.2 mV or ±0.05% of reading whichever is greater
0.00 to 90.00mg/L
±0.2mg/L or ±2% of reading, whichever is greater
0.01, 0.1mg/L
0.0 to 600.0 % saturation
±2 % saturation or ±2% of reading whichever is greater
0.1, 1 % saturation
400 to 850mmHg, automatic using built-in barometer or manual entry
1mmHg
0.0 to 45.0 ppt, automatic correction using manual entry of sample salinity
1 to 5
Water-saturated air, air-saturated water, manual (Winkler titration), zero point
0.1°C, 0.1°F
Manual or automatic with ATC temperature probe
2
Yes, selectable
pH, mV, relative mV (RmV) or ORP with temperature; dissolved oxygen as % saturation or mg/L with temperature and barometric pressure
Transfer single, range or all data points to printer or computer
Automatic data logging with Auto-Read and Timed measure modes; manual data logging with Continuous measure mode
High/low limit alarm, calibration due alarm
English, Spanish, French, Italian, German and Chinese; Portuguese and Korean available with software update
USB, RS232
Universal 50-60 Hz, 100-240 VAC power adapter (Cat. No. 1010003)
800 hours
CE, TUV 3-1, FCC Class A
5° to 45° C (ambient)
24cm x 18cm x 11cm (9.5 in. x 7.1 in. x 4.3 in.)
2 lb.
Promotions
Promotions
  • Thermo Scientific
    Purchase a Thermo Scientific™ Orion Star™ A200 Series Benchtop Meter and receive an Orion Stirring Probe (Cat. No. 13-641-659) — a $260 value — at no additional cost. This is an NLSU exclusive offer. You must apply and qualify for the program to be eligible.
    06-30-2018