Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Orion™ Star A221 pH, mV, ORP and Temperature Portable Waterproof Meter, pH/mV/ORP/Temperature Portable Meter Kit - PROMO

Catalog No. 13645524pm
Click to view available options
Range (pH):
-2 to 20

Make necessary testing easier with the durable and dependable Thermo Scientific™ Orion Star™ A221 pH Portable Meter.

Features:

  • Multi-language interface supports English, Spanish, French, Italian, German and Chinese
  • Thermo Scientific™ AUTO-READ™ locks in the stable reading on your screen and a ready indicator alerts when readings are stable
  • Meter includes up to five point pH calibration with automatic recognition for USA/NIST™ and DIN buffers.
  • Non-volatile memory holds up to 1,000 datapoints with time and date stamp.
  • User can easily transfer data and keep meter software up-to-date with the USB and RS232 ports and complimentary data analysis software.
  • Four AA batteries (included) provide over 800 hours of operation or purchase the universal power adapter (sold separately) to use AC power.
  • Meter is perfectly portable, waterproof and protected to take anywhere with a IP67-rated housing.
TRUSTED_SUSTAINABILITY

Specifications

AC Adapter Universal 50 to 60 Hz, 100 to 240 VAC power adapter (sold separately)
Outputs USB, RS-232
Accuracy (mV) ±0.2 mV or ±0.05% of reading whichever is greater
Accuracy (pH) ±0.002
Alarm Output Calibration due alarm (fixed 12 hour)
Backlight Option Yes (Selectable)
Battery Life 800 hr.
Battery Type 4 x AA
Calibration pH, relative mV (RmV), ORP and temperature
Calibration Points 1 to 5
Certifications/Compliance CE, TUV 3-1, FCC Class A
Data Management Details Transfer single, range or all data points to printer or computer
Data Points 1000 with Date and Time Stamp
Description Orion Star A221 pH portable meter kit with ROSS Ultra Triode low maintenance gel pH/ATC electrode, solutions, meter armor and field case
Channels 1
Display Type LCD Graphic, Backlit
IP Rating IP67
Measurement Modes pH, mV, Relative mV, ORP with Temperature
Range (mV) ±2000 mV
Range (pH) -2 to 20
Resolution (pH) 0.1/0.01/0.001
Temperature Range (Metric) -5.0°C to +105.0°C
Includes Star A221 pH meter, 8107UWMMD ROSS Ultra gel-filled epoxy-body pH/ATC electrode with 3 m cable, pH 4/7/10 buffer and rinse pouches, ROSS storage solution, protective meter armor, hard-sided carrying case, 4 AA batteries, computer cable
Inputs BNC (pH or ORP electrode), pin-tip (reference electrode), 8 pin MiniDIN (ATC temperature probe)
Isopotential Point 7.000 pH
Keypad Comprehensive with menu-specific function keys and dual purpose scroll/shortcut keys
Languages English, Spanish, French, Italian, German and Chinese; Portuguese and Korean available with software update
Log Function Type Automatic with Auto-Read Measure Mode, Manual with Continuous Measure Mode
Memory Non-volatile memory preserves data log, calibration log and meter settings
Methods 10 per channel with password protection
Model Orion Star A221 pH/mV/RmV/ORP/Temperature Portable Meter
Power Supply Universal AC adapter (included) or 4 AA batteries (sold separately)
Probe Type pH electrode, ORP electrode, reference half-cell electrode, ATC temperature probe (includes 8107UWMMD ROSS Ultra Triode pH/ATC electrode)
Range (ORP) ±2000 mV
Range (Relative mV) ±2000 mV
Relative Humidity Range 5 to 85%, non-condensing
Resolution (mV) 0.1 mV
Stability Smart Stability and Smart Averaging
Temperature Accuracy ±0.1°C
Temperature (English) Operating 41°F to 113°F (ambient)
Temperature (Metric) Operating 5°C to 45°C (ambient)
Temperature Probe Calibration 1 point temperature offset calibration
Temperature Range (English) 22.0°F to 221.0° F
Temperature Resolution 0.1°C, 0.1°F
Temperature Selection Manual or automatic with ATC temperature probe
Warranty 3 Year Meter
Dimensions (L x W x H) 24 x 10 x 6.5 cm (9.5 x 3.9 x 2.6 in.)
Weight (English) 1.2 lb.
Weight (Metric) 0.5 kg
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.