Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Orion Star T900 Series Titrator Accessories

Catalog No. 13642990
Click to view available options
Type:
1 L Glass Bottle
1 L Plastic Bottle
10 mL Burette
12 x 1 L Glass Bottles
12 x 1 L Plastic Bottles
20 mL Burette
50 mL Burette
ANZ Power Supply
Burette Cover
Cable Management
China Power Supply
Dispenser Probe
Dispenser Verification Kit
Drying Tube
EU Power Supply
Electrode Holder Assembly
Light-blocking Tubing Kit
Pint Bottle Cap
Reagent Bottle Cap (GL38)
Reagent Bottle Cap (GL45)
Reagent Bottle Holder
Replacement Dispenser Probe
Replacement Reagent Cap
Replacemet Burette
Standard Tubing Kit
Stopper
UK/Singapore Power Supply
US/Japan Power Supply
USB Cable
User Manual
Value Replacement

Minimize maintenance downtime with easy to install Orion Star T900 series titrator replacement parts. Orion Star T900 series titrator accessories expand functionality.

Get accessories and replacement parts for the Orion Star T910 pH titrator, Orion Star T920 redox titrator, Orion Star T930 ion titrator and Orion Star T940 all-in-one titrator. These are designed to expand titrator functionality and minimize maintenance downtime.

•Select interchangable burette sizes of 10mL, 20mL or 50mL to customize your titrator
•Bottle holder helps secure titrant to prevent spills and offers flexible placement
•Color-coded tubing and connectors simplify tubing installation and maintenance
•Computer software is designed to make data transfer simple and reliable

TRUSTED_SUSTAINABILITY

Specifications

Description Orion Star T900 series 240V UK/Singapore power supply
Type UK/Singapore Power Supply
For Use With (Equipment) Orion Star T910 pH titrator, Orion Star T920 redox titrator, Orion Star T930 ion titrator, Orion Star T940 all-in-one titrator
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.