Learn More
Thermo Scientific™ Orion Star T900 Series Titrator Accessories
Description
Get accessories and replacement parts for the Orion Star T910 pH titrator, Orion Star T920 redox titrator, Orion Star T930 ion titrator and Orion Star T940 all-in-one titrator. These are designed to expand titrator functionality and minimize maintenance downtime.
•Select interchangable burette sizes of 10mL, 20mL or 50mL to customize your titrator
•Bottle holder helps secure titrant to prevent spills and offers flexible placement
•Color-coded tubing and connectors simplify tubing installation and maintenance
•Computer software is designed to make data transfer simple and reliable

Specifications
Specifications
Description | Orion Star T900 series light blocking tubing kit, includes one each of burette to valve, titrant cap to valve and dispenser probe to valve tubings with light blocking opacity |
Type | Light-blocking Tubing Kit |
For Use With (Equipment) | Orion Star T910 pH titrator, Orion Star T920 redox titrator, Orion Star T930 ion titrator, Orion Star T940 all-in-one titrator |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.