Learn More
Thermo Scientific™ Orion™ Star A Series Portable Meter Hard-sided Field Case
Description
Use the Thermo Scientific™ Orion™ Star A Series Portable Meter Hard-sided Field Case with the Thermo Scientific™ Orion™ Star A series portable meters (Star A320, Star A220 and Star A120 series) to protect, transport and store your equipment in the field. Field case holds Star A series portable meter (with or without armor), portable electrode holders, extra batteries, solutions, pH electrode in storage sleeve, conductivity probe and dissolved oxygen probe in storage sleeve. The electrodes and probes can remain connected to meter while in the case plus the internal separating door can be removed for cable management.
Hard-sided field case includes cable management system for electrodes.
Specifications
Specifications
| Description | Orion Star A series hard-sided field case |
| Type | Meter Accessories |
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.