Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ORMDL1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA566786
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA566786 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA566786 Supplier Invitrogen™ Supplier No. PA566786
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: FETPDQGKARL LTHWEQLDYG VQFTSSR Highest antigen sequence identity to the following orthologs - mouse 96%, rat 93%.

The ORMDL (ORM1-like) family of proteins consist of ORMDL1, ORMDL2 and ORMDL3 that may play roles in -gamma-secretase complex maturation. Localized to the membrane of the endoplasmic reticulum, ORMDLs are widely expressed and are implicated in the expression of Presenilin (PSI) which is involved in Alzheimer's disease (AD). Expression of ORMDL1 is down-regulated in PSI mutations, suggesting a possible role as a therapeutic target for AD. ORMDLs have recently been identified as a potential risk factor for childhood asthma.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ORMDL1
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ORMDL1
Gene Accession No. Q9P0S3
Gene Alias Adoplin-1; C730042F17Rik; HSPC202; ORM1-like 1; ORM1-like 1 (S. cerevisiae); ORM1-like protein 1; ORMDL sphingolipid biosynthesis regulator 1; ORMDL1
Gene Symbols ORMDL1
Host Species Rabbit
Immunogen Recombinant Human ORMDL1. Recombinant protein control fragment (Product #RP-105362).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 94101
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.