Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ORMDL3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA524232
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA524232 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA524232 Supplier Invitrogen™ Supplier No. PA524232
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This antibody is predicted to react with bovine, rat and Xenopus and zebrafish based on sequence homology.

Negative regulator of sphingolipid synthesis. May indirectly regulate endoplasmic reticulum-mediated Ca(+2) signaling.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ORMDL3
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene ORMDL3
Gene Accession No. Q8N138, Q9CPZ6
Gene Alias 2810011N17Rik; AI315223; ORM1-like 3 (S. cerevisiae); ORM1-like protein 3; ORMDL sphingolipid biosynthesis regulator 3; Ormdl3
Gene Symbols ORMDL3
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 53-81 amino acids from the central region of human ORMDL3.
Purification Method Antigen affinity chromatography, Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 66612, 94103
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.