Learn More
Invitrogen™ ORMDL3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA590144
Description
Immunogen sequence: MNVGTAHSEV NPNTRVMNSR GIWLSYVLAI GLLHIVLLSI PFVSVPVVWT LTNLIHNMGM YIFLHTVKGT PFETPDQGKA RLLTHWEQMD YGVQFTASRK; Positive Samples: 293T, Mouse eye, Mouse thymus, Mouse liver; Cellular Location: Endoplasmic reticulum membrane, Multi-pass membrane protein.
Negative regulator of sphingolipid synthesis. May indirectly regulate endoplasmic reticulum-mediated Ca(+2) signaling.
Specifications
ORMDL3 | |
Polyclonal | |
Unconjugated | |
ORMDL3 | |
2810011N17Rik; AI315223; ORM1-like 3 (S. cerevisiae); ORM1-like protein 3; ORMDL sphingolipid biosynthesis regulator 3; Ormdl3 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
66612, 94103 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot | |
1.19 mg/mL | |
PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
Q8N138, Q9CPZ6 | |
ORMDL3 | |
A synthetic peptide corresponding to a sequence within amino acids 1-100 of human ORMDL3 (NP_6448091). | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.