Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ornithine Carbamoyltransferase Antibody (CL4049), Novus Biologicals™

Mouse Monoclonal Antibody
$382.00 - $610.00
Specifications
Antigen | Ornithine Carbamoyltransferase |
---|---|
Applications | Western Blot |
Classification | Monoclonal |
Conjugate | Unconjugated |
Form | Purified |
Description
Ornithine Carbamoyltransferase Monoclonal specifically detects Ornithine Carbamoyltransferase in Human samples. It is validated for Western Blot.Specifications
Ornithine Carbamoyltransferase | |
Monoclonal | |
Purified | |
RUO | |
Human | |
5009 | |
This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:ILADYLTLQEHYSSLKGLTLSWIGDGNNILHSIMMSAAKFGMHLQAATPKGYEPDASVTKLAEQYAKENGTKLLLTNDPLEAAHGGNVLITDTWISMGQEEEKKKRLQAFQGYQVTMKTAKVAASDWT | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
Western Blot | |
Unconjugated | |
Mouse | |
metabolism | |
EC 2.1.3, EC 2.1.3.3, MGC129967, MGC129968, MGC138856, OCTD, ornithine carbamoyltransferase, ornithine carbamoyltransferase, mitochondrial, Ornithine transcarbamylase, OTCase | |
OTC | |
IgG1 | |
Protein A purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title