Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ornithine Carbamoyltransferase Antibody (OTI8A1), DyLight 680, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP273149FR
Description
Ornithine Carbamoyltransferase Monoclonal specifically detects Ornithine Carbamoyltransferase in Human, Mouse, Rat samples. It is validated for Western Blot.Specifications
Ornithine Carbamoyltransferase | |
Monoclonal | |
DyLight 680 | |
PBS with 0.05% Sodium Azide | |
OTC | |
Recombinant protein fragment corresponding to amino acids 33-354 of human Ornithine Carbamoyltransferase (NP_000522)produced in E.coli. | |
0.1 mL | |
Primary | |
Human, Mouse, Rat | |
IgG1 |
Western Blot | |
OTI8A1 | |
Western Blot | |
EC 2.1.3, EC 2.1.3.3, MGC129967, MGC129968, MGC138856, OCTD, ornithine carbamoyltransferase, ornithine carbamoyltransferase, mitochondrial, Ornithine transcarbamylase, OTCase | |
Mouse | |
Affinity Purified | |
metabolism | |
5009.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction