Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ornithine Decarboxylase Antibody (ODC1/3636R), FITC, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP308792F
Description
Ornithine Decarboxylase Monoclonal antibody specifically detects Ornithine Decarboxylase in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry (Paraffin)Specifications
Ornithine Decarboxylase | |
Monoclonal | |
FITC | |
ODCEC 4.1.1.17, ornithine decarboxylase, ornithine decarboxylase 1 | |
Recombinant full-length human Ornithine Decarboxylase protein (Uniprot: P11926) | |
100 μg | |
Cell Cycle and Replication | |
4953 | |
Store at 4°C in the dark. | |
IgG |
Western Blot, Immunohistochemistry (Paraffin) | |
ODC1/3636R | |
PBS | |
Rabbit | |
Protein A or G purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction