Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ornithine Decarboxylase Antibody (rODC1/487), Alexa Fluor™ 405, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP314027AF405
This item is not returnable.
View return policy
Description
Ornithine Decarboxylase Monoclonal antibody specifically detects Ornithine Decarboxylase in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, ImmunofluorescenceSpecifications
Ornithine Decarboxylase | |
Monoclonal | |
Alexa Fluor 405 | |
ODCEC 4.1.1.17, ornithine decarboxylase, ornithine decarboxylase 1 | |
Recombinant full-length human Ornithine Decarboxylase protein (Uniprot: P11926) | |
0.1 mL | |
Cell Cycle and Replication | |
4953 | |
Store at 4°C in the dark. | |
IgG1 κ |
Western Blot, ELISA, Immunofluorescence | |
rODC1/487 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction