Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ornithine Decarboxylase Antibody (SR1904), Novus Biologicals™

Rabbit Monoclonal Antibody
$302.50 - $413.50
Specifications
Antigen | Ornithine Decarboxylase |
---|---|
Clone | SR1904 |
Dilution | Western Blot 1:500-1:2000 |
Applications | Western Blot |
Classification | Monoclonal |
Description
Ornithine Decarboxylase Monoclonal antibody specifically detects Ornithine Decarboxylase in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
Ornithine Decarboxylase | |
Western Blot 1:500-1:2000 | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse, Rat | |
ODCEC 4.1.1.17, ornithine decarboxylase, ornithine decarboxylase 1 | |
A synthesized peptide derived from human Ornithine Decarboxylase (Uniprot #: P11926) | |
Primary | |
Store at -20°C. Avoid freeze/thaw cycles. |
SR1904 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Cell Cycle and Replication | |
PBS (pH 7.4), 150 mM NaCl, 50% Glycerol | |
4953 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title