Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ORNT1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA569271
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA569271 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA569271 Supplier Invitrogen™ Supplier No. PA569271
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This target displays homology in the following species: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 93%; Human: 100%; Mouse: 100%; Rabbit: 86%; Rat: 100%; Sheep: 93%.

The SLC25A15 gene encodes the mitochondrial ornithine transporter, which transports ornithine across the inner mitochondrial membrane from the cytosol to the mitochondrial matrix. This is a vital step in the urea cycle, which serves to eliminate toxic ammonium ions from the breakdown of nitrogen.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ORNT1
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene SLC25A15
Gene Accession No. Q9WVD5
Gene Alias Ab1-114; D13S327; D630044L02Rik; HHH; MGC137403 protein; mitochondrial ornithine transporter 1; ORC1; ornithine transporter 1; ornithine transporter) member 15; ornithine transporter, mitochondrial 1; Ornt1; SLC25A15; solute carrier family 25 (mitochondrial carrier ornithine transporter), member 15; solute carrier family 25 (mitochondrial carrier; ornithine transporter) member 15; solute carrier family 25 member 15; SP1855
Gene Symbols SLC25A15
Host Species Rabbit
Immunogen synthetic peptide directed towards the middle region of human ORNT1 (aa 20-69).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 18408
Target Species Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.