Learn More
Invitrogen™ ORNT1 Polyclonal Antibody

Description
ORNT1 Polyclonal Antibody for Western Blot, IHC (P), ELISA
Specifications
Specifications
| Antigen | ORNT1 |
| Applications | ELISA, Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 0.48 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | SLC25A15 |
| Gene Accession No. | Q9WVD5, Q9Y619 |
| Gene Alias | Ab1-114; D13S327; D630044L02Rik; HHH; MGC137403 protein; mitochondrial ornithine transporter 1; ORC1; ornithine transporter 1; ornithine transporter) member 15; ornithine transporter, mitochondrial 1; Ornt1; SLC25A15; solute carrier family 25 (mitochondrial carrier ornithine transporter), member 15; solute carrier family 25 (mitochondrial carrier; ornithine transporter) member 15; solute carrier family 25 member 15; SP1855 |
| Gene Symbols | SLC25A15 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.