Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ORNT1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA528976
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA528976 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA528976 Supplier Invitrogen™ Supplier No. PA528976
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: 293T, A431, Jurkat, Raji. Predicted reactivity: Mouse (95%), Rat (84%), Pig (97%), Chicken (81%), Sheep (94%), Bovine (95%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

The SLC25A15 gene encodes the mitochondrial ornithine transporter, which transports ornithine across the inner mitochondrial membrane from the cytosol to the mitochondrial matrix. This is a vital step in the urea cycle, which serves to eliminate toxic ammonium ions from the breakdown of nitrogen.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ORNT1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.71 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7
Gene SLC25A15
Gene Accession No. Q9WVD5, Q9Y619
Gene Alias Ab1-114; D13S327; D630044L02Rik; HHH; MGC137403 protein; mitochondrial ornithine transporter 1; ORC1; ornithine transporter 1; ornithine transporter) member 15; ornithine transporter, mitochondrial 1; Ornt1; SLC25A15; solute carrier family 25 (mitochondrial carrier ornithine transporter), member 15; solute carrier family 25 (mitochondrial carrier; ornithine transporter) member 15; solute carrier family 25 member 15; SP1855
Gene Symbols SLC25A15
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human SLC25A15. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10166, 18408
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.