Learn More
Stoelting™ Orofacial Pain Assessment Device
SDP
Stoelting™ Orofacial Pain Assessment Device provides an automated measurement of hot or cold or mechanical orofacial pain assessment in the trigeminal nerve in rats and mice. Using an operant behavior conflict paradigm, rodents voluntarily decide between receiving a reward or avoiding an aversive stimulus.
$8080.00 - $14360.00
Specifications
For Use With (Application) Neuroscience
Animal Mouse, Rat
View More Specs
Products 2
Catalog Number Mfr. No. Model Price Quantity & Availability  
Catalog Number Mfr. No. Model Price Quantity & Availability  
10-000-376
SDP
View Documents
Stoelting™
67500
OPAD system
Each for $14,360.00
Only null left
Add to Cart
10-000-377
SDP
View Documents
Stoelting™
67501
OPAD Additional cage
Each for $8,080.00
Only null left
Add to Cart
Description
Description
Specifications
Specifications
Neuroscience
Mouse, Rat
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.