Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Orosomucoid 2 Antibody (959445), Alexa Fluor™ 350, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB9306U
Description
Orosomucoid 2 Monoclonal antibody specifically detects Orosomucoid 2 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, CyTOFSpecifications
Orosomucoid 2 | |
Monoclonal | |
Alexa Fluor 350 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
Primary | |
Human | |
Purified |
Flow Cytometry, Immunocytochemistry, CyTOF | |
959445 | |
Intracellular Staining by Flow Cytometry, Immunocytochemistry, CyTOF-ready | |
AGP 2, AGP2, AGP2alpha-1-acid glycoprotein 2, AGP-B, AGP-B', alpha-1-acid glycoprotein, type 2, OMD 2, orosomucoid 2, orosomucoid-2 | |
Chinese hamster ovary cell line CHO-derived recombinant human ORM-2, Met1-Glu201, Accession # P19652 | |
0.1 mL | |
Cardiovascular Biology, Immunology | |
5005 | |
Store at 4C in the dark. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction