Learn More
ORP Standard 600 mV vs. Ag/AgCl, Reagents
Specifications
Specifications
| Chemical Name or Material | ORP Standard 600 mV vs. Ag/AgCl |
| Packaging | Glass Bottle |
| CAS | 7732-18-5, 7783-85-9, 7664-93-9 |
| Specific Gravity | 1.0286 |
| Physical Form | Liquid |
| Color | Amber |
| Boiling Point | 100°C |
| For Use With (Equipment) | Redox Meter |
| Synonym | Oxidation Reduction Potential Standard |
| Solubility | Completely miscible |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.