Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ORP1 Polyclonal Antibody
Rabbit Polyclonal Antibody
$589.50
Specifications
Antigen | ORP1 |
---|---|
Content And Storage | Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. |
Applications | Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
PIPA5111129
|
Invitrogen™
PA5111129 |
100 μL |
Each for $589.50
|
|
|||||
PIPA5111175
|
Invitrogen™
PA5111175 |
100 μL |
Each for $589.50
|
|
|||||
Description
Immunogen sequence: VPKNSLQQSR EDWLEAIEEH SAYSTHYCSQ DQLTDEEEED TVSAADLKKS LEKAQSCQQR LDREISNFLK MIKECDMAKE MLPSFLQKVE VVSEA.
This gene encodes a member of the oxysterol-binding protein family, a group of intracellular lipid receptors. Most members contain an N-terminal pleckstrin homology domain and a highly conserved C-terminal OSBP-like sterol-binding domain, although some members contain only the sterol-binding domain. Transcript variants derived from alternative promoter usage and/or alternative splicing exist; they encode different isoforms.Specifications
ORP1 | |
Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rabbit | |
RUO | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9BXW6 | |
114876 | |
Recombinant Protein corresonding to Human ORP1. Recombinant protein control fragment (Product #RP-98278). | |
Antibody |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Polyclonal | |
Liquid | |
IgG | |
Human | |
OSBPL1A | |
DCDC4A; FLJ10217; FLJ50293; FLJ55454; FLJ79410; G430090F17Rik; Gm753; ORP1; ORP-1; Orp1a; Orp1l; OSBP8; OSBPL1; OSBPL1A; OSBPL1B; OSBP-related protein 1; oxysterol binding protein like 1A; oxysterol binding protein-like 1A; oxysterol binding protein-like 1B; oxysterol-binding protein-related protein 1; oxysterol-binding protein-related protein 1 variant 1; oxysterol-binding protein-related protein 1 variant 2 | |
OSBPL1A | |
Primary | |
Antigen affinity chromatography |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title