Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ORP150/HSP12A Antibody (201), mFluor Violet 450 SE, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP289890MFV450
Description
ORP150/HSP12A Monoclonal antibody specifically detects ORP150/HSP12A in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin)Specifications
ORP150/HSP12A | |
Monoclonal | |
mFluor Violet 450 SE | |
50mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Western Blot, ELISA, Immunocytochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
201 | |
Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin | |
150 kDa oxygen-regulated protein, 170 kDa glucose-regulated protein, DKFZp686N08236, FLJ97572, glucose-regulated protein 170, GRP170, GRP-170, HSP12A, hypoxia up-regulated 1, hypoxia up-regulated protein 1, ORP-150, ORP150FLJ94899, oxygen regulated protein (150kD) | |
Rabbit immunized with purified, recombinant Human ORP150/HSP12A (Accession#: NP_001124463.1, Met 695-Leu 994) | |
0.1 mL | |
Hypoxia | |
10525 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction