Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ORP2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA583621
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA583621 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA583621 Supplier Invitrogen™ Supplier No. PA583621
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: VEGHIQDKNK KKLFMIYGKW TECLWGIDPV SYESFKKQER RGDHLRKAKL DEDSGKADSD VADDVPVAQE TVQVIPGSKL Highest antigen sequence indentity to the following orthologs: Mouse - 79%, Rat - 80%.

This gene encodes a member of the oxysterol-binding protein (OSBP) family, a group of intracellular lipid receptors. Most members contain an N-terminal pleckstrin homology domain and a highly conserved C-terminal OSBP-like sterol-binding domain, although some members contain only the sterol-binding domain. This encoded protein contains only the sterol-binding domain. In vitro studies have shown that the encoded protein can bind strongly to phosphatic acid and weakly to phosphatidylinositol 3-phosphate, but cannot bind to 25-hydroxycholesterol. The protein associates with the Golgi apparatus. Transcript variants encoding different isoforms have been described.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ORP2
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.4 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene OSBPL2
Gene Accession No. Q9H1P3
Gene Alias C130070J12Rik; DFNA67; DNFA67; FLJ20223; KIAA0772; MGC4307; MGC8342; mKIAA0772; ORP2; ORP-2; Osbpl2; OSBP-related protein 2; oxysterol binding protein like 2; oxysterol binding protein-like 2; oxysterol-binding protein-like 2; oxysterol-binding protein-related protein 2; RP11-157P1.2
Gene Symbols OSBPL2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human OSBPL2. Recombinant protein control fragment (Product #RP-99418).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9885
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.