Learn More
Invitrogen™ ORP2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA589675
Description
Immunogen sequence: NKKKLFMIYG KWTECLWGID PVSYESFKKQ ERRGDHLRKA KLDEDSGKAD SDVADDVPVA QETVQVIPGS KLLWRINTRP PNSAQMYNFT SFTVSLNELE TGMEKTLPPT DCRLRPDIRG MENGNMDLAS QEKERLEEKQ REARRERAKE EAEWQTRWFY PGNNPYTGTP DWLYAGDYFE RNFSDCPDIY; Positive Samples: U-251MG, A549, HepG2, HeLa.
This gene encodes a member of the oxysterol-binding protein (OSBP) family, a group of intracellular lipid receptors. Most members contain an N-terminal pleckstrin homology domain and a highly conserved C-terminal OSBP-like sterol-binding domain, although some members contain only the sterol-binding domain. This encoded protein contains only the sterol-binding domain. In vitro studies have shown that the encoded protein can bind strongly to phosphatic acid and weakly to phosphatidylinositol 3-phosphate, but cannot bind to 25-hydroxycholesterol. The protein associates with the Golgi apparatus. Transcript variants encoding different isoforms have been described.
Specifications
ORP2 | |
Polyclonal | |
Unconjugated | |
OSBPL2 | |
C130070J12Rik; DFNA67; DNFA67; FLJ20223; KIAA0772; MGC4307; MGC8342; mKIAA0772; ORP2; ORP-2; Osbpl2; OSBP-related protein 2; oxysterol binding protein like 2; oxysterol binding protein-like 2; oxysterol-binding protein-like 2; oxysterol-binding protein-related protein 2; RP11-157P1.2 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
9885 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot | |
1.01 mg/mL | |
PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
Q9H1P3 | |
OSBPL2 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 291-480 of human OSBPL2 (NP_6530811). | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.