Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ORP7 Monoclonal Antibody (3D11)
GREENER_CHOICE

Catalog No. PIMA524509
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA524509 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA524509 Supplier Invitrogen™ Supplier No. MA524509
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Peptide Sequence: ELNELTAELK RSLPSTDTRL RPDQRYLEEG NIQAAEAQKR RIEQLQRDRR KVMEENNIVH QARFFRRQTD SSGKEWWVTN NTYWRLRAEP GYGNMDGAVL W.

This gene encodes a member of the oxysterol-binding protein (OSBP) family, a group of intracellular lipid receptors. Like most members, the encoded protein contains an N-terminal pleckstrin homology domain and a highly conserved C-terminal OSBP-like sterol-binding domain. Two transcript variants encoding the same isoform have been identified.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ORP7
Applications ELISA, Western Blot
Classification Monoclonal
Clone 3D11
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7.4
Gene OSBPL7
Gene Accession No. Q9BZF2
Gene Alias ORP7; ORP-7; OSBPL7; OSBP-related protein 7; oxysterol binding protein like 7; oxysterol binding protein-like 7; oxysterol-binding protein-related protein 7
Gene Symbols OSBPL7
Host Species Mouse
Immunogen OSBPL7 (NP_665741.1, 742 a.a. ~ 842 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 114881
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.