Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ORP7 Polyclonal Antibody

Goat Polyclonal Antibody
Supplier: Invitrogen™ PA5142079
Description
This antibody is tested in Peptide ELISA: antibody detection limit dilution 1:32,000.
This gene encodes a member of the oxysterol-binding protein (OSBP) family, a group of intracellular lipid receptors. Like most members, the encoded protein contains an N-terminal pleckstrin homology domain and a highly conserved C-terminal OSBP-like sterol-binding domain. Two transcript variants encoding the same isoform have been identified.
Specifications
ORP7 | |
Polyclonal | |
Unconjugated | |
OSBPL7 | |
ORP7; ORP-7; OSBPL7; OSBP-related protein 7; oxysterol binding protein like 7; oxysterol binding protein-like 7; oxysterol-binding protein-related protein 7 | |
Goat | |
Ammonium Sulfate Precipitation | |
RUO | |
114881 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunocytochemistry | |
0.5 mg/mL | |
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
Q9BZF2 | |
OSBPL7 | |
Peptide with sequence C-EPGYGNMDGAVLW. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction