Learn More
Invitrogen™ ORP8 Polyclonal Antibody

Description
Immunogen sequence: LDPLTGEWHY KFADTRPWDP LNDMIQFEKD GVIQTKVKHR TPMVSVPKMK HKPTRQQKKV AKGYSSPEPD IQDSSGSEAQ SVKPSTRRKK GIELGDIQSS IESIKQTQEE IKRNIMALRN HLVSSTPATD Y Highest antigen sequence identity to the following orthologs: Mouse - 97%, Rat - 97%.
Specifications
Specifications
| Antigen | ORP8 |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Concentration | 0.1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | OSBPL8 |
| Gene Accession No. | Q9BZF1 |
| Gene Alias | AA536976; AA536995; C730029P18Rik; D330025H14Rik; Kiaa1451; MST120; MSTP120; ORP8; ORP-8; OSBP10; Osbpl8; OSBP-related protein 8; oxysterol binding protein like 8; oxysterol binding protein-like 8; oxysterol-binding protein-related protein 8; RGD1561474 |
| Gene Symbols | OSBPL8 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.