Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ORP8 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5110065
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5110065 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5110065 Supplier Invitrogen™ Supplier No. PA5110065
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ORP8 Polyclonal Antibody for ELISA, IHC (P), Western Blot

This gene encodes a member of the oxysterol-binding protein (OSBP) family, a group of intracellular lipid receptors. Like most members, the encoded protein contains an N-terminal pleckstrin homology domain and a highly conserved C-terminal OSBP-like sterol-binding domain. Two transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ORP8
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 2.24 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene OSBPL8
Gene Accession No. B9EJ86, Q9BZF1
Gene Alias AA536976; AA536995; C730029P18Rik; D330025H14Rik; Kiaa1451; MST120; MSTP120; ORP8; ORP-8; OSBP10; Osbpl8; OSBP-related protein 8; oxysterol binding protein like 8; oxysterol binding protein-like 8; oxysterol-binding protein-related protein 8; RGD1561474
Gene Symbols OSBPL8
Host Species Rabbit
Immunogen Recombinant protein (or fragment).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 314824
Target Species Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.