Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ORP8 Polyclonal Antibody

Rabbit Polyclonal Antibody

Supplier:  Invitrogen™ PA531833

Catalog No. PIPA531833


Only null left
Add to Cart

Description

Description

Recommended positive controls: U87-MG. Predicted reactivity: Mouse (96%), Rat (97%), Rhesus Monkey (100%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

This gene encodes a member of the oxysterol-binding protein (OSBP) family, a group of intracellular lipid receptors. Like most members, the encoded protein contains an N-terminal pleckstrin homology domain and a highly conserved C-terminal OSBP-like sterol-binding domain. Two transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

ORP8
Polyclonal
Unconjugated
OSBPL8
AA536976; AA536995; C730029P18Rik; D330025H14Rik; Kiaa1451; MST120; MSTP120; ORP8; ORP-8; OSBP10; Osbpl8; OSBP-related protein 8; oxysterol binding protein like 8; oxysterol binding protein-like 8; oxysterol-binding protein-related protein 8; RGD1561474
Rabbit
Antigen affinity chromatography
RUO
114882
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Western Blot, Immunocytochemistry
1 mg/mL
0.1M Tris glycine with 20% glycerol and 0.01% thimerosal; pH 7
Q9BZF1
OSBPL8
Recombinant fragment corresponding to a region within amino acids 463 and 773 of Human ORP8.
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

WARNING: Cancer - www.P65Warnings.ca.gov
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.