Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ORP9 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA582972
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA582972 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA582972 Supplier Invitrogen™ Supplier No. PA582972
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: QGLDSGFVPS VQDFDKKLTE ADAYLQILIE QLKLFDDKLQ NCKEDEQRKK IETLKETTNS MVESIKHCIV LLQIAKDQSN AEKHADGM Highest antigen sequence indentity to the following orthologs: Mouse - 93%, Rat - 83%.

This gene encodes a member of a subfamily of LIM domain proteins that are characterized by an N-terminal proline rich region and three C-terminal LIM domains. The encoded protein localizes to the cell periphery in focal adhesions and may be involved in cell-cell adhesion and cell motility. This protein also shuttles through the nucleus and may function as a transcriptional co-activator. This gene is located at the junction of certain disease related chromosomal translocations which result in the expression of chimeric proteins that may promote tumor growth. Alternate splicing results in multiple transcript variants.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ORP9
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene OSBPL9
Gene Accession No. Q96SU4
Gene Alias 2600011I06Rik; AU015843; FLJ12492; FLJ14629; FLJ14801; FLJ32055; FLJ34384; MGC15035; Orp9; ORP-9; OSBP4; OSBPL9; OSBP-related protein 9; oxysterol binding protein like 9; oxysterol binding protein-like 9; oxysterol-binding protein-like protein 9; Oxysterol-binding protein-like protein 9-like protein; oxysterol-binding protein-related protein 9; RP4-657D16.1; Unknown (protein for MGC:140169); zgc:154069
Gene Symbols OSBPL9
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human OSBPL9. Recombinant protein control fragment (Product #RP-94136).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 114883
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.