Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ORP9 Polyclonal Antibody
Goat Polyclonal Antibody
Supplier: Invitrogen™ PA518222
Description
This antibody is predicted to react with bovine, canine, mouse, porcine and rat based on sequence homology. This antibody is tested in Peptide ELISA: antibody detection limit dilution 32,000.
This gene encodes a member of a subfamily of LIM domain proteins that are characterized by an N-terminal proline rich region and three C-terminal LIM domains. The encoded protein localizes to the cell periphery in focal adhesions and may be involved in cell-cell adhesion and cell motility. This protein also shuttles through the nucleus and may function as a transcriptional co-activator. This gene is located at the junction of certain disease related chromosomal translocations which result in the expression of chimeric proteins that may promote tumor growth. Alternate splicing results in multiple transcript variants.
Specifications
ORP9 | |
Polyclonal | |
Unconjugated | |
OSBPL9 | |
2600011I06Rik; AU015843; FLJ12492; FLJ14629; FLJ14801; FLJ32055; FLJ34384; MGC15035; Orp9; ORP-9; OSBP4; OSBPL9; OSBP-related protein 9; oxysterol binding protein like 9; oxysterol binding protein-like 9; oxysterol-binding protein-like protein 9; Oxysterol-binding protein-like protein 9-like protein; oxysterol-binding protein-related protein 9; RP4-657D16.1; Unknown (protein for MGC:140169); zgc:154069 | |
Goat | |
Ammonium Sulfate Precipitation | |
RUO | |
114883 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.5 mg/mL | |
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
Q96SU4 | |
OSBPL9 | |
Synthetic peptide sequence (EPLLKRLGAAKH) corresponding to the C-terminus amino acids of OSBPL9 (aa 725-736). | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction