Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Orphenadrine Citrate, USP, 98-101.5%, Spectrum™ Chemical
SDP

Catalog No. 18604907
Click to view available options
Quantity:
5 g
25 g
100 g
1 kg
25 kg
Packaging:
Amber Glass Bottle
Fiber Drum

O1047, 4682-36-4, C18H23NO·C6H8O7

Spectrum™ Chemical Orphenadrine Citrate, USP , an anticholinergic drug, has been used to treat painful muscle spasms. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 4682-36-4
Molecular Formula C24H31NO8
Molecular Weight (g/mol) 461.51
InChI Key MMMNTDFSPSQXJP-UHFFFAOYNA-N
IUPAC Name 2-hydroxypropane-1,2,3-tricarboxylic acid; dimethyl({2-[(2-methylphenyl)(phenyl)methoxy]ethyl})amine
SMILES OC(=O)CC(O)(CC(O)=O)C(O)=O.CN(C)CCOC(C1=CC=CC=C1)C1=CC=CC=C1C

Specifications

CAS Max % 1
Ignition Residue 0.001
Loss on Drying 0.005
Packaging Amber Glass Bottle
Quantity 5 g
Percent Purity 98 to 101.5%
Grade USP
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.