Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
QuidelOrtho BioClone™ Reagents

Click to view available options
Quantity:
1 x 5 mL
3 x 10 mL
15 x 10 mL
Type:
Anti-C (Anti-RH2) BioClone™ Reagent
Anti-D BioClone™ Reagent
Anti-E (Anti-RH3) BioClone™ Reagent
Anti-c BioClone™ Reagent
Anti-e BioClone™ Reagent
Description
Anti Sera products provide qualitative tests for recognition of antigens on human red blood cells
- Using high-protein reagents prepared from human serum, Anti-D blood grouping reagent recognizes D (Rh0) antigens
- Anti-C (Anti-RH2), Anti-E (Anti-RH3), Anti-c (Anti-RH4) and Anti-e (Anti-RH5) blood grouping reagents are low-protein reagents prepared from human/mouse cell lines; used to recognize corresponding antigens on human red blood cells
- The control is prepared from pools of human serum tested and found to be negative for blood group antibodies
Specifications
Specifications
Type | Anti-D BioClone™ Reagent |
For Use With (Application) | To recognize spontaneous Agglutination of red Blood cells |
Quantity | 15 x 10 mL |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction