Learn More
QuidelOrtho BioClone™ Reagents
Click to view available options
Quantity:
1 x 5 mL
3 x 10 mL
15 x 10 mL
Type:
Anti-C (Anti-RH2) BioClone™ Reagent
Anti-D BioClone™ Reagent
Anti-E (Anti-RH3) BioClone™ Reagent
Anti-c BioClone™ Reagent
Anti-e BioClone™ Reagent
Description
Anti Sera products provide qualitative tests for recognition of antigens on human red blood cells
- Using high-protein reagents prepared from human serum, Anti-D blood grouping reagent recognizes D (Rh0) antigens
- Anti-C (Anti-RH2), Anti-E (Anti-RH3), Anti-c (Anti-RH4) and Anti-e (Anti-RH5) blood grouping reagents are low-protein reagents prepared from human/mouse cell lines; used to recognize corresponding antigens on human red blood cells
- The control is prepared from pools of human serum tested and found to be negative for blood group antibodies
Specifications
Specifications
Type | Anti-C (Anti-RH2) BioClone™ Reagent |
For Use With (Application) | To recognize spontaneous Agglutination of red Blood cells |
Quantity | 1 x 5 mL |