Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
QuidelOrtho RESOLVE™ Reagent Red Blood Cells (Panels): Panel A

Click to view available options
Type:
3% Resolve™ Panel A
8% Resolve™ Panel A
Description
- Prepared in 3% (for tube tests) and 0.8% (for ID-MTS™ brand Gel Test™) suspensions
RESOLVE Panel A:
- 11-cell panel that provides selection of cells to assure identification of antibodies to common blood group factors, including those known to exhibit dosag
- Cell 11 will represent unusual Rh phenotype, possess antigen of low incidence or lack antigen of high incidence
Specifications
Specifications
| Type | 3% Resolve™ Panel A |
| Quantity | 11 x 3 mL |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction