Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

QuidelOrtho VITROS™ 5600 Integrated System

Catalog No. 23057105
Change view
Click to view available options
Capacity (Metric):
2 to 80 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
23-057-105 2 to 80 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is a non-standard voltage; please ensure you are ordering the correct product. Please contact customer service for assistance: 1-800-766-7000.
Catalog No. 23-057-105 Supplier QuidelOrtho Supplier No. 6802413
Only null left
Add to Cart
Add to Cart
Request a Quote
This item is a non-standard voltage; please ensure you are ordering the correct product. Please contact customer service for assistance: 1-800-766-7000.

Specifications

Type Vitros™ 5600 Integrated System
Sample Type Serum, Plasma, Urine, CSF, Amniotic Fluid, Whole Blood
Analysis Time Potentiometric ≈2.5 min., colorimetric ≈6 min., immuno rate ≈8 min., MicroTip™ ≈8 to 16 min., microwell ≈16 to 73 min., for single result
Sample Size 80 samples in universal sample trays, 10 samples in dedicated STAT lane
Humidity 15 to 75%
Operating Temperature 15°C to 30°C
Calibration 84 Days
Capacity (Metric) 2 to 80 μL
Includes Color coded graphical user interface, ergonomic flat, low-glare, LCD, 17 in. touchscreen monitor, numeric keypad on monitor, detached keyboard for maximum flexibility, onboard documentation and animated help (V-Docs), onboard maintenance documentation
Depth (English) 33.5 in.
Depth (Metric) 85.09 cm
Height (English) 64.5 in.
Height (Metric) 1.64 m
Width (English) 83.5 in.
Width (Metric) 212.09 cm
Electrical Requirements 200 to 240V 47 to 63Hz
Amperage 30 A
Frequency 47/63 Hz
Voltage 200/240 V
Weight (English) 1580 lb.
Weight (Metric) 717 kg
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.