Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ortho-Phosphoric acid 85%, Suprapur™, MilliporeSigma™

Catalog No. M1005520500 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
250 mL
500 mL
1 L
2.5 L
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M1005520500 500 mL
M1005520250 250 mL
M1005521000 1 L
M1005522500 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. M1005520500 Supplier MilliporeSigma Supplier No. 1.00552.0500
Add to Cart
Add to Cart

Chemical Identifiers

CAS 7664-38-2
Molecular Formula H3O4P
InChI Key NBIIXXVUZAFLBC-UHFFFAOYSA-N
IUPAC Name phosphoric acid

Specifications

pH < 0.5 (100 g/L, H2O, 20°C)
Melting Point 21°C
Boiling Point 158°C (1013 mbar)
Density 1.71 g/cm3 (at 20°C)
Packaging Plastic Bottle
Vapor Pressure 2 hPa (20°C)
Assay Percent Range ≥ 85.0%
Quantity 500 mL
Viscosity 30.5 mm2/s (20°C)
Synonym o-Phosphoric acid, Orthophosphoric acid
SMILES OP(O)(O)=O
Molecular Weight (g/mol) 97.99
Show More Show Less
Recommended Storage Store above 15°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.