Learn More
QuidelOrtho Reagent Red Cells
Click to view available options
Quantity:
1 x 10 mL
10 mL
2 x 10 mL
3 x 10 mL
4 x 5 mL
Each
Type:
Affirmagen™
Affirmagen™ 3 Reagent Red Cells
Affirmagen™ Reagent Red Cells
Alba Q-Chek™ Simulated Whole Blood Control
Coombs Control Reagent Red Cells
Fetalscreen™ II
Pooled Screening Reagent Red Cells
SelectogeN™ Reagent Red Cells
Surgiscreen™ Reagent Red Cells
Description
- Tests include hemagglutination and hemolysis-based results
- A variety of testing functions are available, including:
- Detection and/or identification of unexpected blood group antibodies
- Affirmation of expected group antibodies
- Serum grouping
- Confirmation of negative antiglobulin tests
- Screening for RhD (Rh0) positive fetal red blood cells
Specifications
Specifications
Type | Alba Q-Chek™ Simulated Whole Blood Control |
For Use With (Application) | For detection and identification of a variety of Blood group Antibodies |
Quantity | Each |