Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Orthophenanthroline TS, 0.025 M Phenanthroline Ferrous Sulfate, Ricca Chemical
SDP

Catalog No. 5464816 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
120 mL
500 mL
Packaging:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
54648-16 500 mL Amber Glass Bottle
54648-4 120 mL Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 54648-16 Supplier Ricca Chemical Company Supplier No. 5464.816
Add to Cart
Add to Cart

Chemical Identifiers

CAS 7732-18-5 , 5144-89-8 , 7782-63-0
Synonym dihydrogen oxide, dihydrogen monoxide

Specifications

Color Red
CAS Min % 96.0, 1.44, 0.67
CAS Max % 100.0, 1.5, 0.7
Grade Laboratory
Concentration or Composition (by Analyte or Components) 0.025M Phenanthroline Ferrous Sulfate
Packaging Amber Glass Bottle
Quantity 500 mL
Physical Form Liquid
Chemical Name or Material Orthophenanthroline TS
ShelfLife 24 months
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.