Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
OS9 Antibody, mFluor Violet 450 SE, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB100519MFV450
Description
OS9 Polyclonal antibody specifically detects OS9 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), KnockDownSpecifications
OS9 | |
Polyclonal | |
Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Knockdown Validated | |
Amplified in osteosarcoma 9, endoplasmic reticulum lectin 2, ERLEC2, erlectin 2, OS-9, osteosarcoma amplified 9, endoplasmic reticulum associated protein, osteosarcoma amplified 9, endoplasmic reticulum lectin, protein OS-9 | |
Synthetic peptide made to an internal portion of isoform 1 of the human OS9 protein (within residues 300-400). [Swiss-Prot# Q13438] | |
0.1 mL | |
Cancer, Chromatin Research, Hypoxia, Immunology | |
10956 | |
Store at 4C in the dark. | |
IgG |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin), KnockDown | |
mFluor Violet 450 SE | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human, Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction