Learn More
Invitrogen™ OS9 Polyclonal Antibody

Description
In Western blot, bands can be seen at ~75 kDa representing isoform 1 and at ~70 kDa representing isoform 2. Suggested positive control: HEK293 postnuclear extracts, antigen standard for OS9 (transient overexpression lysate).
Specifications
Specifications
| Antigen | OS9 |
| Applications | Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | Phosphate, tris citrate with 0.01% sodium azide; pH 7-8 |
| Gene | OS9 |
| Gene Accession No. | Q13438, Q8K2C7 |
| Gene Alias | 4632413K17Rik; amplified in osteosarcoma; amplified in osteosarcoma 9; AU022351; endoplasmic reticulum lectin 2; ERLEC2; erlectin 2; OS9; Os-9; OS9 endoplasmic reticulum lectin; OS9, endoplasmic reticulum lectin; osteosarcoma amplified 9; osteosarcoma amplified 9, endoplasmic reticulum associated protein; osteosarcoma amplified 9, endoplasmic reticulum lectin; protein OS-9 |
| Gene Symbols | OS9 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.