Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ OS9 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA553078
Description
Immunogen sequence: PLSCSYVLTI RTPRLCPHPL LRPPPSAAPQ AILCHPSLQP EEYMAYVQRQ ADSKQYGDKI IEELQDLGPQ VWSETKSGVA PQKMAGASPT KDDSKDSDFW KML Highest antigen sequence identity to the following orthologs: Mouse - 66%, Rat - 70%.
OS9 is an ER glycoprotein found in the intralumenal level and highly expressed in tumor tissue. The OS9 gene is transcriptionally induced upon activation of the Ire1/Xbp1 ER-stress pathway.
Specifications
OS9 | |
Polyclonal | |
Unconjugated | |
OS9 | |
4632413K17Rik; amplified in osteosarcoma; amplified in osteosarcoma 9; AU022351; endoplasmic reticulum lectin 2; ERLEC2; erlectin 2; OS9; Os-9; OS9 endoplasmic reticulum lectin; OS9, endoplasmic reticulum lectin; osteosarcoma amplified 9; osteosarcoma amplified 9, endoplasmic reticulum associated protein; osteosarcoma amplified 9, endoplasmic reticulum lectin; protein OS-9 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
10956 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q13438 | |
OS9 | |
Recombinant protein corresponding to Human OS9. Recombinant protein control fragment (Product #RP-90253). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction