Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ OS9 Recombinant Rabbit Monoclonal Antibody (26H3)
GREENER_CHOICE

Catalog No. FEMA550013
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEMA550013 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEMA550013 Supplier Invitrogen™ Supplier No. MA550013
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

OS9 Recombinant Monoclonal Antibody for Western Blot, IHC (P), ELISA

OS9 is an ER glycoprotein found in the intralumenal level and highly expressed in tumor tissue. The OS9 gene is transcriptionally induced upon activation of the Ire1/Xbp1 ER-stress pathway.
TRUSTED_SUSTAINABILITY

Specifications

Antigen OS9
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Recombinant Monoclonal
Clone 26H3
Concentration 0.75 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene OS9
Gene Accession No. Q13438, Q8K2C7
Gene Alias 4632413K17Rik; amplified in osteosarcoma; amplified in osteosarcoma 9; AU022351; endoplasmic reticulum lectin 2; ERLEC2; erlectin 2; OS9; Os-9; OS9 endoplasmic reticulum lectin; OS9, endoplasmic reticulum lectin; osteosarcoma amplified 9; osteosarcoma amplified 9, endoplasmic reticulum associated protein; osteosarcoma amplified 9, endoplasmic reticulum lectin; protein OS-9
Gene Symbols OS9
Host Species Rabbit
Immunogen A synthesized peptide derived from human OS9.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10956, 216440
Target Species Human, Mouse
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.